The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145392.1 | NP_001138864.1 | 2 - 3 | 322 | 81 |
NM_001145393.2 | NP_001138865.1 | 2 - 3 | 290 | 81 | |
NM_001145394.1 | NP_001138866.1 | 2 - 3 | 290 | 81 | |
NM_001321277.1 | NP_001308206.1 | 2 - 3 | 290 | 81 | |
NM_001321278.1 | NP_001308207.1 | 2 - 3 | 290 | 81 | |
NM_001321279.1 | NP_001308208.1 | 2 - 3 | 556 | 81 | |
NM_025265.3 | NP_079541.1 | 2 - 3 | 574 | 81 | |
NR_135607.1 | 2 - 3 | 290 | 81 | ||
XM_017007292.1 | XP_016862781.1 | 2 - 3 | 242 | 81 | |
XM_017007293.1 | XP_016862782.1 | 2 - 3 | 574 | 81 | |
XM_017007294.1 | XP_016862783.1 | 2 - 3 | 308 | 81 | |
XM_017007295.1 | XP_016862784.1 | 2 - 3 | 504 | 81 | |
XM_017007296.1 | XP_016862785.1 | 2 - 3 | 870 | 81 | |
XM_017007297.1 | XP_016862786.1 | 2 - 3 | 574 | 81 | |
GenBank mRNA | AK027821.1 | - | 2 - 3 | 574 | 81 |
AK074794.1 | - | 2 - 3 | 221 | 81 | |
AK225875.1 | - | 2 - 3 | 279 | 81 | |
AK291183.1 | - | 2 - 3 | 359 | 81 | |
AK300449.1 | - | 2 - 3 | 539 | 81 | |
BC004178.2 | - | 2 - 3 | 307 | 81 | |
BC004211.2 | - | 2 - 3 | 307 | 81 | |
BC019582.1 | - | 2 - 3 | 556 | 81 | |
DA614108.1 | - | 2 - 3 | 291 | 81 |