The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145392.1 | NP_001138864.1 | 4 - 5 | 444 | 116 |
NM_001145393.2 | NP_001138865.1 | 4 - 5 | 412 | 116 | |
NM_001145394.1 | NP_001138866.1 | 4 - 5 | 412 | 116 | |
NM_001321277.1 | NP_001308206.1 | 4 - 5 | 412 | 116 | |
NM_001321278.1 | NP_001308207.1 | 4 - 5 | 412 | 116 | |
NM_001321279.1 | NP_001308208.1 | 4 - 5 | 678 | 116 | |
NM_025265.3 | NP_079541.1 | 4 - 5 | 696 | 116 | |
NR_135607.1 | 4 - 5 | 412 | 116 | ||
XM_017007296.1 | XP_016862785.1 | 4 - 5 | 992 | 116 | |
GenBank mRNA | AK027821.1 | - | 4 - 5 | 696 | 116 |
AK074794.1 | - | 4 - 5 | 343 | 116 | |
AK225875.1 | - | 4 - 5 | 401 | 116 | |
AK291183.1 | - | 4 - 5 | 481 | 116 | |
AK300449.1 | - | 4 - 5 | 661 | 116 | |
BC004178.2 | - | 4 - 5 | 429 | 116 | |
BC004211.2 | - | 4 - 5 | 429 | 116 | |
BC019582.1 | - | 4 - 5 | 678 | 116 | |
CN341716.1 | - | 3 - 4 | 149 | 116 | |
DA614108.1 | - | 4 - 5 | 413 | 116 |