Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSEN2
Entrez Gene ID
80746
Gene Name
tRNA splicing endonuclease subunit 2
Gene Aliases
PCH2B, SEN2, SEN2L
Chromosome Location
Chr.3: 12484432 - 12539624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145392.1 NP_001138864.1 5 - 6 973 102
NM_001145394.1 NP_001138866.1 6 - 7 764 102
NM_001321277.1 NP_001308206.1 5 - 6 941 102
NM_001321278.1 NP_001308207.1 5 - 6 941 102
NM_025265.3 NP_079541.1 5 - 6 1225 102
NR_135607.1 5 - 6 941 102
XM_011534139.2 XP_011532441.1 3 - 4 1124 102
XM_017007292.1 XP_016862781.1 6 - 7 950 102
XM_017007293.1 XP_016862782.1 6 - 7 1282 102
XM_017007294.1 XP_016862783.1 6 - 7 1016 102
XM_017007295.1 XP_016862784.1 6 - 7 1212 102
XM_017007296.1 XP_016862785.1 5 - 6 1521 102
GenBank mRNA AK225875.1 - 6 - 7 753 102
AK291183.1 - 5 - 6 1010 102
AK295508.1 - 4 - 5 1100 102
BC004178.2 - 5 - 6 958 102
BC004211.2 - 5 - 6 958 102
BC019582.1 - 5 - 6 1207 102
BC021975.2 - 1 - 2 208 102

View Full Product Details