The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145392.1 | NP_001138864.1 | 6 - 7 | 1040 | 117 |
NM_001145394.1 | NP_001138866.1 | 7 - 8 | 831 | 117 | |
NM_001321277.1 | NP_001308206.1 | 6 - 7 | 1008 | 117 | |
NM_001321278.1 | NP_001308207.1 | 6 - 7 | 1008 | 117 | |
NM_025265.3 | NP_079541.1 | 6 - 7 | 1292 | 117 | |
NR_135607.1 | 6 - 7 | 1008 | 117 | ||
XM_011534139.2 | XP_011532441.1 | 4 - 5 | 1191 | 117 | |
XM_017007292.1 | XP_016862781.1 | 7 - 8 | 1017 | 117 | |
XM_017007293.1 | XP_016862782.1 | 7 - 8 | 1349 | 117 | |
XM_017007294.1 | XP_016862783.1 | 7 - 8 | 1083 | 117 | |
XM_017007295.1 | XP_016862784.1 | 7 - 8 | 1279 | 117 | |
XM_017007296.1 | XP_016862785.1 | 6 - 7 | 1588 | 117 | |
GenBank mRNA | AK225875.1 | - | 7 - 8 | 820 | 117 |
AK291183.1 | - | 6 - 7 | 1077 | 117 | |
AK295508.1 | - | 5 - 6 | 1167 | 117 | |
BC004178.2 | - | 6 - 7 | 1025 | 117 | |
BC004211.2 | - | 6 - 7 | 1025 | 117 | |
BC019582.1 | - | 6 - 7 | 1274 | 117 | |
BC021975.2 | - | 2 - 3 | 275 | 117 |