Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYH10
Entrez Gene ID
4628
Gene Name
myosin, heavy chain 10, non-muscle
Gene Aliases
NMMHC-IIB, NMMHCB
Chromosome Location
Chr.17: 8474205 - 8631369 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256012.1 NP_001242941.1 14 - 15 1745 86
NM_001256095.1 NP_001243024.1 14 - 15 1742 86
NM_005964.3 NP_005955.3 13 - 14 1715 86
XM_011523875.2 XP_011522177.1 14 - 15 2034 86
XM_011523878.2 XP_011522180.1 14 - 15 2086 86
XM_011523879.2 XP_011522181.1 14 - 15 2031 86
XM_011523880.2 XP_011522182.1 13 - 14 2004 86
XM_017024677.1 XP_016880166.1 14 - 15 1800 86
XM_017024678.1 XP_016880167.1 15 - 16 1782 86
XM_017024679.1 XP_016880168.1 15 - 16 1823 86
XM_017024680.1 XP_016880169.1 15 - 16 2131 86
XM_017024681.1 XP_016880170.1 14 - 15 1752 86
XM_017024682.1 XP_016880171.1 14 - 15 1727 86
GenBank mRNA AB210026.1 - 14 - 15 1697 86
BC117690.1 - 12 - 13 1617 86
BC117691.1 - 12 - 13 1617 86
BC144668.1 - 14 - 15 1699 86
BC150634.1 - 14 - 15 1699 86
M69181.1 - 13 - 14 1655 86

View Full Product Details