Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASCC2
Entrez Gene ID
84164
Gene Name
activating signal cointegrator 1 complex subunit 2
Gene Aliases
ASC1p100, p100
Chromosome Location
Chr.22: 29788608 - 29838304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032204.4 NP_115580.2 3 - 4 384 62
XM_005261775.2 XP_005261832.1 4 - 5 424 62
XM_011530442.2 XP_011528744.1 4 - 5 424 62
XM_011530443.2 XP_011528745.1 5 - 6 490 62
XM_011530444.2 XP_011528746.1 5 - 6 537 62
XM_011530445.2 XP_011528747.1 4 - 5 471 62
XM_011530446.2 XP_011528748.1 3 - 4 354 62
XM_011530450.2 XP_011528752.1 5 - 6 540 62
XM_011530451.2 XP_011528753.1 6 - 7 614 62
XM_011530452.2 XP_011528754.1 5 - 6 587 62
XM_011530453.2 XP_011528755.1 4 - 5 442 62
XM_011530454.2 XP_011528756.1 3 - 4 326 62
XM_011530455.2 XP_011528757.1 2 - 3 255 62
XM_017028991.1 XP_016884480.1 3 - 4 2562 62
XM_017028992.1 XP_016884481.1 3 - 4 2562 62
XM_017028993.1 XP_016884482.1 3 - 4 2562 62
XM_017028994.1 XP_016884483.1 3 - 4 2562 62
XM_017028996.1 XP_016884485.1 4 - 5 1364 62
XM_017028998.1 XP_016884487.1 4 - 5 1364 62
XM_017028999.1 XP_016884488.1 5 - 6 490 62
XM_017029000.1 XP_016884489.1 4 - 5 471 62
XM_017029005.1 XP_016884494.1 5 - 6 540 62
XM_017029006.1 XP_016884495.1 6 - 7 614 62
XM_017029007.1 XP_016884496.1 4 - 5 442 62
XM_017029008.1 XP_016884497.1 3 - 4 326 62
XM_017029009.1 XP_016884498.1 2 - 3 255 62
GenBank mRNA AK022886.1 - 5 - 6 502 62
AY013289.1 - 2 - 3 261 62
BC025368.2 - 3 - 4 345 62

View Full Product Details