Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASCC2
Entrez Gene ID
84164
Gene Name
activating signal cointegrator 1 complex subunit 2
Gene Aliases
ASC1p100, p100
Chromosome Location
Chr.22: 29788608 - 29838304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032204.4 NP_115580.2 4 - 5 554 116
XM_005261775.2 XP_005261832.1 5 - 6 594 116
XM_011530442.2 XP_011528744.1 5 - 6 594 116
XM_011530443.2 XP_011528745.1 6 - 7 660 116
XM_011530444.2 XP_011528746.1 6 - 7 707 116
XM_011530445.2 XP_011528747.1 5 - 6 641 116
XM_011530446.2 XP_011528748.1 4 - 5 524 116
XM_011530448.2 XP_011528750.1 4 - 5 427 116
XM_011530449.2 XP_011528751.1 3 - 4 365 116
XM_011530450.2 XP_011528752.1 6 - 7 710 116
XM_011530451.2 XP_011528753.1 7 - 8 784 116
XM_011530452.2 XP_011528754.1 6 - 7 757 116
XM_011530453.2 XP_011528755.1 5 - 6 612 116
XM_011530454.2 XP_011528756.1 4 - 5 496 116
XM_011530455.2 XP_011528757.1 3 - 4 425 116
XM_017028991.1 XP_016884480.1 4 - 5 2732 116
XM_017028992.1 XP_016884481.1 4 - 5 2732 116
XM_017028993.1 XP_016884482.1 4 - 5 2732 116
XM_017028994.1 XP_016884483.1 4 - 5 2732 116
XM_017028995.1 XP_016884484.1 3 - 4 2573 116
XM_017028996.1 XP_016884485.1 5 - 6 1534 116
XM_017028997.1 XP_016884486.1 3 - 4 2573 116
XM_017028998.1 XP_016884487.1 5 - 6 1534 116
XM_017028999.1 XP_016884488.1 6 - 7 660 116
XM_017029000.1 XP_016884489.1 5 - 6 641 116
XM_017029001.1 XP_016884490.1 4 - 5 427 116
XM_017029002.1 XP_016884491.1 3 - 4 365 116
XM_017029003.1 XP_016884492.1 5 - 6 501 116
XM_017029004.1 XP_016884493.1 3 - 4 365 116
XM_017029005.1 XP_016884494.1 6 - 7 710 116
XM_017029006.1 XP_016884495.1 7 - 8 784 116
XM_017029007.1 XP_016884496.1 5 - 6 612 116
XM_017029008.1 XP_016884497.1 4 - 5 496 116
XM_017029009.1 XP_016884498.1 3 - 4 425 116
GenBank mRNA AK022886.1 - 6 - 7 672 116
AY013289.1 - 3 - 4 431 116
BC025368.2 - 4 - 5 515 116
BP201867.1 - 3 - 4 401 116

View Full Product Details