The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317824.1 | NP_001304753.1 | 3 - 4 | 494 | 122 |
NM_001317825.1 | NP_001304754.1 | 3 - 4 | 585 | 122 | |
NM_001317826.1 | NP_001304755.1 | 4 - 5 | 557 | 122 | |
NM_017872.4 | NP_060342.2 | 3 - 4 | 589 | 122 | |
XM_017009603.1 | XP_016865092.1 | 3 - 4 | 524 | 122 | |
GenBank mRNA | AK000553.1 | - | 3 - 4 | 565 | 122 |
AK021663.1 | - | 3 - 4 | 540 | 122 | |
AK023355.1 | - | 2 - 3 | 203 | 122 | |
AK223119.1 | - | 3 - 4 | 558 | 122 | |
AK304590.1 | - | 3 - 4 | 465 | 122 | |
AL136669.1 | - | 3 - 4 | 557 | 122 | |
AY463216.1 | - | 3 - 4 | 663 | 122 | |
BC001523.2 | - | 3 - 4 | 535 | 122 | |
BC001852.2 | - | 3 - 4 | 535 | 122 | |
BC023521.2 | - | 3 - 4 | 555 | 122 | |
CR533503.1 | - | 3 - 4 | 537 | 122 |