Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C17orf53
Entrez Gene ID
78995
Gene Name
chromosome 17 open reading frame 53
Gene Aliases
-
Chromosome Location
Chr.17: 44141902 - 44162477 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171251.2 NP_001164722.1 7 - 8 1882 87
NM_001321310.1 NP_001308239.1 5 - 6 1657 87
NM_001321311.1 NP_001308240.1 7 - 8 1879 87
NM_024032.4 NP_076937.2 7 - 8 1885 87
XM_011525187.1 XP_011523489.1 7 - 8 1732 87
XM_011525188.1 XP_011523490.1 6 - 7 1651 87
XM_011525189.1 XP_011523491.1 6 - 7 1594 87
XM_011525190.1 XP_011523492.1 6 - 7 1591 87
XM_011525191.2 XP_011523493.1 7 - 8 1735 87
XM_011525193.1 XP_011523495.1 5 - 6 1507 87
XM_011525197.2 XP_011523499.1 4 - 5 900 87
XM_017025031.1 XP_016880520.1 7 - 8 1908 87
XM_017025032.1 XP_016880521.1 7 - 8 1904 87
GenBank mRNA AK291924.1 - 7 - 8 1876 87
AL834295.1 - 7 - 8 1829 87
BC000121.1 - 5 - 6 1537 87

View Full Product Details