Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCA8
Entrez Gene ID
10351
Gene Name
ATP binding cassette subfamily A member 8
Gene Aliases
-
Chromosome Location
Chr.17: 68867285 - 68955426 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288985.1 NP_001275914.1 10 - 11 1613 88
NM_001288986.1 NP_001275915.1 9 - 10 1452 88
NM_007168.3 NP_009099.1 9 - 10 1452 88
XM_005256938.2 XP_005256995.1 9 - 10 1480 88
XM_005256940.4 XP_005256997.1 8 - 9 1385 88
XM_011524191.2 XP_011522493.1 9 - 10 1443 88
XM_011524192.2 XP_011522494.1 9 - 10 1443 88
GenBank mRNA AB020629.1 - 9 - 10 1411 88
AK295876.1 - 9 - 10 1425 88
AK304286.1 - 8 - 9 1284 88
AK304360.1 - 9 - 10 1423 88
BC037309.1 - 10 - 11 1381 88
BC130280.1 - 10 - 11 1462 88

View Full Product Details