The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288985.1 | NP_001275914.1 | 26 - 27 | 3772 | 148 |
NM_001288986.1 | NP_001275915.1 | 25 - 26 | 3611 | 148 | |
NM_007168.3 | NP_009099.1 | 24 - 25 | 3491 | 148 | |
XM_005256938.2 | XP_005256995.1 | 25 - 26 | 3639 | 148 | |
XM_005256940.4 | XP_005256997.1 | 24 - 25 | 3544 | 148 | |
GenBank mRNA | AB020629.1 | - | 24 - 25 | 3450 | 148 |
BC130280.1 | - | 26 - 27 | 3621 | 148 |