The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016001.2 | NP_057085.2 | 9 - 10 | 1252 | 91 |
XM_006721930.3 | XP_006721993.2 | 9 - 10 | 1231 | 91 | |
XM_011524870.2 | XP_011523172.1 | 9 - 10 | 1231 | 91 | |
GenBank mRNA | AF151806.1 | - | 9 - 10 | 1238 | 91 |
AK026355.1 | - | 10 - 11 | 1001 | 91 | |
AK314399.1 | - | 9 - 10 | 1231 | 91 | |
AY007138.1 | - | 7 - 8 | 722 | 91 | |
BC025276.1 | - | 9 - 10 | 1228 | 91 | |
BM548967.1 | - | 5 - 6 | 91 |