Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CDKL3
- Entrez Gene ID
-
51265
- Gene Name
- cyclin dependent kinase like 3
- Gene Aliases
-
NKIAMRE
- Chromosome Location
-
Chr.5: 134298424 - 134371159 on Build GRCh38
View Full Product Details