Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDKL3
Entrez Gene ID
51265
Gene Name
cyclin dependent kinase like 3
Gene Aliases
NKIAMRE
Chromosome Location
Chr.5: 134298424 - 134371159 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113575.1 NP_001107047.1 10 - 11 1570 115
NM_001300853.1 NP_001287782.1 9 - 10 1336 115
XM_011543429.2 XP_011541731.1 10 - 11 1921 115
XM_011543439.2 XP_011541741.1 7 - 8 1074 115
XM_011543444.2 XP_011541746.1 6 - 7 903 115
XM_011543445.2 XP_011541747.1 6 - 7 852 115
XM_011543447.2 XP_011541749.1 5 - 6 763 115
XM_017009524.1 XP_016865013.1 10 - 11 2398 115
XM_017009525.1 XP_016865014.1 11 - 12 1966 115
XM_017009526.1 XP_016865015.1 11 - 12 1964 115
XM_017009527.1 XP_016865016.1 10 - 11 2362 115
XM_017009528.1 XP_016865017.1 9 - 10 2353 115
XM_017009529.1 XP_016865018.1 10 - 11 2398 115
XM_017009530.1 XP_016865019.1 10 - 11 2398 115
XM_017009531.1 XP_016865020.1 10 - 11 2398 115
XM_017009532.1 XP_016865021.1 9 - 10 2316 115
XM_017009533.1 XP_016865022.1 9 - 10 2352 115
XM_017009534.1 XP_016865023.1 9 - 10 2352 115
XM_017009535.1 XP_016865024.1 9 - 10 2316 115
XM_017009536.1 XP_016865025.1 7 - 8 2111 115
XM_017009537.1 XP_016865026.1 8 - 9 1516 115
XM_017009538.1 XP_016865027.1 10 - 11 1406 115
XM_017009539.1 XP_016865028.1 9 - 10 1227 115
XM_017009540.1 XP_016865029.1 8 - 9 1037 115
XM_017009541.1 XP_016865030.1 9 - 10 1563 115
XM_017009542.1 XP_016865031.1 8 - 9 994 115
XM_017009543.1 XP_016865032.1 8 - 9 1021 115
XM_017009544.1 XP_016865033.1 8 - 9 1272 115
XM_017009545.1 XP_016865034.1 7 - 8 1079 115
XM_017009546.1 XP_016865035.1 7 - 8 1108 115
XM_017009547.1 XP_016865036.1 5 - 6 800 115
GenBank mRNA AK294559.1 - 7 - 8 958 115
AK294740.1 - 6 - 7 868 115
AK299305.1 - 6 - 7 818 115
AK301802.1 - 9 - 10 1330 115
AK310955.1 - 6 - 7 808 115
BC041799.1 - 10 - 11 1572 115

View Full Product Details