The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172698.1 | NP_001166169.1 | 3 - 4 | 560 | 90 |
NM_001172699.1 | NP_001166170.1 | 3 - 4 | 591 | 90 | |
XM_011537555.2 | XP_011535857.1 | 3 - 4 | 502 | 90 | |
XM_011537556.2 | XP_011535858.1 | 3 - 4 | 1850 | 90 | |
GenBank mRNA | AF468497.1 | - | 3 - 4 | 514 | 90 |
AK123614.1 | - | 3 - 4 | 591 | 90 | |
BC144251.1 | - | 3 - 4 | 560 | 90 |