Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL17RC
Entrez Gene ID
84818
Gene Name
interleukin 17 receptor C
Gene Aliases
CANDF9, IL17-RL, IL17RL
Chromosome Location
Chr.3: 9917074 - 9933627 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203263.1 NP_001190192.1 15 - 16 1575 77
NM_001203264.1 NP_001190193.1 14 - 15 1524 77
NM_001203265.1 NP_001190194.1 13 - 14 1479 77
NM_032732.5 NP_116121.2 14 - 15 1530 77
NM_153460.3 NP_703190.1 15 - 16 1575 77
NM_153461.3 NP_703191.1 15 - 16 1764 77
NR_037807.1 13 - 14 1741 77
XM_017007326.1 XP_016862815.1 15 - 16 1767 77
XM_017007327.1 XP_016862816.1 15 - 16 1767 77
XM_017007328.1 XP_016862817.1 15 - 16 1728 77
XM_017007329.1 XP_016862818.1 15 - 16 1764 77
XM_017007330.1 XP_016862819.1 15 - 16 1725 77
XM_017007331.1 XP_016862820.1 14 - 15 1722 77
XM_017007332.1 XP_016862821.1 14 - 15 1719 77
XM_017007333.1 XP_016862822.1 14 - 15 1713 77
XM_017007334.1 XP_016862823.1 14 - 15 1692 77
XM_017007335.1 XP_016862824.1 14 - 15 1680 77
XM_017007336.1 XP_016862825.1 14 - 15 1713 77
XM_017007337.1 XP_016862826.1 13 - 14 1647 77
XM_017007338.1 XP_016862827.1 14 - 15 1680 77
XM_017007339.1 XP_016862828.1 13 - 14 1668 77
XM_017007340.1 XP_016862829.1 15 - 16 1555 77
XM_017007341.1 XP_016862830.1 14 - 15 1510 77
XM_017007342.1 XP_016862831.1 14 - 15 1479 77
XM_017007343.1 XP_016862832.1 14 - 15 1476 77
XM_017007344.1 XP_016862833.1 15 - 16 1513 77
XM_017007345.1 XP_016862834.1 14 - 15 1471 77
XM_017007346.1 XP_016862835.1 14 - 15 1507 77
XM_017007347.1 XP_016862836.1 13 - 14 1434 77
XM_017007348.1 XP_016862837.1 13 - 14 1417 77
XM_017007349.1 XP_016862838.1 13 - 14 1431 77
GenBank mRNA AF458065.1 - 15 - 16 1530 77
AK092907.1 - 15 - 16 1765 77
AK094746.1 - 3 - 4 1046 77
AK225650.1 - 14 - 15 1469 77
AK303171.1 - 12 - 13 1290 77
AK315024.1 - 13 - 14 1718 77
AK315053.1 - 14 - 15 1475 77
AY358840.1 - 14 - 15 1492 77
AY359098.1 - 14 - 15 1520 77
EF676032.1 - 13 - 14 1390 77
EF676033.1 - 15 - 16 1333 77
EF676034.1 - 14 - 15 1293 77

View Full Product Details