Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C10orf71
Entrez Gene ID
118461
Gene Name
chromosome 10 open reading frame 71
Gene Aliases
-
Chromosome Location
Chr.10: 49297009 - 49330145 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135196.1 NP_001128668.1 3 - 3 3752 59
XM_005269476.4 XP_005269533.1 3 - 3 3817 59
XM_005269477.3 XP_005269534.1 3 - 3 3737 59
XM_005269478.4 XP_005269535.1 3 - 3 3805 59
XM_017015619.1 XP_016871108.1 4 - 4 3527 59
GenBank mRNA AJ311357.1 - 1 - 1 838 59
AK056949.1 - 1 - 1 660 59
AL833265.1 - 3 - 3 3701 59

View Full Product Details