Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMCO5A
Entrez Gene ID
145942
Gene Name
transmembrane and coiled-coil domains 5A
Gene Aliases
TMCO5
Chromosome Location
Chr.15: 37934607 - 37997541 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152453.3 NP_689666.2 8 - 9 698 87
XM_005254168.3 XP_005254225.1 8 - 9 679 87
XM_011521262.2 XP_011519564.1 8 - 9 679 87
XM_011521263.1 XP_011519565.1 8 - 9 679 87
XM_011521264.1 XP_011519566.1 8 - 9 679 87
GenBank mRNA AK093126.1 - 8 - 9 679 87
AK310517.1 - 8 - 9 684 87
BC029221.2 - 7 - 8 611 87
BI826726.1 - 8 - 9 662 87

View Full Product Details