Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AP1M1
Entrez Gene ID
8907
Gene Name
adaptor related protein complex 1 mu 1 subunit
Gene Aliases
AP47, CLAPM2, CLTNM, MU-1A
Chromosome Location
Chr.19: 16197854 - 16235345 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130524.1 NP_001123996.1 3 - 4 434 68
NM_032493.3 NP_115882.1 3 - 4 434 68
XM_017027420.1 XP_016882909.1 3 - 4 374 68
XM_017027421.1 XP_016882910.1 3 - 4 374 68
GenBank mRNA AB209808.1 - 3 - 4 354 68
AF290613.1 - 3 - 4 337 68
AK027528.1 - 3 - 4 342 68
AK297824.1 - 3 - 4 377 68
BC017469.2 - 3 - 4 379 68
DC386056.1 - 3 - 4 434 68
DQ059565.1 - 3 - 4 261 68

View Full Product Details