The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130065.1 | NP_001123537.1 | 19 - 20 | 2842 | 79 |
NM_004145.3 | NP_004136.2 | 19 - 20 | 2842 | 79 | |
GenBank mRNA | AB209997.1 | - | 19 - 20 | 2837 | 79 |
AB290184.1 | - | 19 - 20 | 2837 | 79 |