The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130065.1 | NP_001123537.1 | 25 - 26 | 4523 | 85 |
NM_004145.3 | NP_004136.2 | 25 - 26 | 4523 | 85 | |
GenBank mRNA | AB209997.1 | - | 25 - 26 | 4518 | 85 |
AB290184.1 | - | 25 - 26 | 4518 | 85 | |
AY927576.1 | - | 2 - 3 | 324 | 85 |