The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130065.1 | NP_001123537.1 | 9 - 10 | 1685 | 59 |
NM_004145.3 | NP_004136.2 | 9 - 10 | 1685 | 59 | |
GenBank mRNA | AB209997.1 | - | 9 - 10 | 1680 | 59 |
AB290184.1 | - | 9 - 10 | 1680 | 59 |