Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHMP5
Entrez Gene ID
51510
Gene Name
charged multivesicular body protein 5
Gene Aliases
C9orf83, CGI-34, HSPC177, PNAS-2, SNF7DC2, Vps60
Chromosome Location
Chr.9: 33264879 - 33282069 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016410.5 NP_057494.3 6 - 7 689 64
GenBank mRNA AF132968.1 - 6 - 7 517 64
AF161525.1 - 6 - 7 509 64
AF229832.1 - 3 - 4 251 64
AF275810.1 - 3 - 4 251 64
AK024753.1 - 6 - 7 537 64
AK315455.1 - 6 - 7 538 64
BC006974.1 - 6 - 7 519 64
BC007457.1 - 6 - 7 551 64
BC016698.1 - 6 - 7 568 64
BC018603.1 - 6 - 7 530 64
BC020796.1 - 6 - 7 520 64
BC021168.1 - 6 - 7 534 64

View Full Product Details