Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NBEA
Entrez Gene ID
26960
Gene Name
neurobeachin
Gene Aliases
BCL8B, LYST2
Chromosome Location
Chr.13: 34942287 - 35672737 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015678.4 NP_056493.3 39 - 40 6842 110
XM_005266346.4 XP_005266403.1 39 - 40 6842 110
XM_005266347.4 XP_005266404.2 39 - 40 6842 110
XM_005266348.4 XP_005266405.2 38 - 39 6833 110
XM_006719803.3 XP_006719866.1 39 - 40 6842 110
XM_006719805.3 XP_006719868.1 38 - 39 6833 110
XM_006719806.3 XP_006719869.1 38 - 39 6833 110
XM_011535046.2 XP_011533348.1 31 - 32 4973 110
XM_011535047.2 XP_011533349.1 39 - 40 6842 110
XM_017020544.1 XP_016876033.1 39 - 40 6824 110
XM_017020545.1 XP_016876034.1 39 - 40 6824 110
XM_017020546.1 XP_016876035.1 39 - 40 6130 110
GenBank mRNA AB046764.1 - 6 - 7 554 110
AF467288.1 - 40 - 41 6514 110
AK091469.1 - 10 - 11 1001 110
AK122888.1 - 9 - 10 1355 110
AK127791.1 - 16 - 17 2193 110

View Full Product Details