Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NBEA
Entrez Gene ID
26960
Gene Name
neurobeachin
Gene Aliases
BCL8B, LYST2
Chromosome Location
Chr.13: 34942287 - 35672737 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204197.1 NP_001191126.1 7 - 8 796 86
NM_015678.4 NP_056493.3 47 - 48 7826 86
XM_005266346.4 XP_005266403.1 47 - 48 7826 86
XM_005266347.4 XP_005266404.2 47 - 48 7826 86
XM_005266348.4 XP_005266405.2 46 - 47 7817 86
XM_006719803.3 XP_006719866.1 47 - 48 7826 86
XM_006719805.3 XP_006719868.1 46 - 47 7817 86
XM_006719806.3 XP_006719869.1 46 - 47 7817 86
XM_011535046.2 XP_011533348.1 39 - 40 5957 86
XM_017020544.1 XP_016876033.1 47 - 48 7808 86
XM_017020545.1 XP_016876034.1 47 - 48 7808 86
XM_017020546.1 XP_016876035.1 47 - 48 7114 86
GenBank mRNA AB046764.1 - 14 - 15 1538 86
AF467288.1 - 48 - 49 7498 86
AK091469.1 - 18 - 19 1985 86
AK122888.1 - 17 - 18 2339 86
AK127791.1 - 24 - 25 3177 86
AK294737.1 - 7 - 8 796 86
AK297773.1 - 8 - 9 895 86
AK315939.1 - 7 - 8 793 86
AL137748.1 - 1 - 2 438 86
BX640760.1 - 23 - 24 3156 86

View Full Product Details