Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NBEA
Entrez Gene ID
26960
Gene Name
neurobeachin
Gene Aliases
BCL8B, LYST2
Chromosome Location
Chr.13: 34942287 - 35672737 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204197.1 NP_001191126.1 11 - 12 1398 87
NM_015678.4 NP_056493.3 51 - 52 8428 87
XM_005266346.4 XP_005266403.1 51 - 52 8413 87
XM_005266347.4 XP_005266404.2 52 - 53 8491 87
XM_005266348.4 XP_005266405.2 51 - 52 8482 87
XM_006719803.3 XP_006719866.1 52 - 53 8476 87
XM_006719805.3 XP_006719868.1 50 - 51 8404 87
XM_006719806.3 XP_006719869.1 50 - 51 8419 87
XM_011535046.2 XP_011533348.1 44 - 45 6622 87
XM_017020544.1 XP_016876033.1 52 - 53 8473 87
XM_017020545.1 XP_016876034.1 51 - 52 8410 87
XM_017020546.1 XP_016876035.1 52 - 53 7779 87
GenBank mRNA AB046764.1 - 18 - 19 2140 87
AF072371.1 - 5 - 6 615 87
AF467288.1 - 52 - 53 8100 87
AK091469.1 - 22 - 23 2587 87
AK122888.1 - 21 - 22 2941 87
AK294737.1 - 11 - 12 1398 87
AK297773.1 - 12 - 13 1496 87
AL137748.1 - 6 - 7 1103 87
BX640760.1 - 27 - 28 3758 87

View Full Product Details