The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015678.4 | NP_056493.3 | 8 - 9 | 1765 | 106 |
XM_005266346.4 | XP_005266403.1 | 8 - 9 | 1765 | 106 | |
XM_005266347.4 | XP_005266404.2 | 8 - 9 | 1765 | 106 | |
XM_005266348.4 | XP_005266405.2 | 8 - 9 | 1765 | 106 | |
XM_006719803.3 | XP_006719866.1 | 8 - 9 | 1765 | 106 | |
XM_006719805.3 | XP_006719868.1 | 8 - 9 | 1765 | 106 | |
XM_006719806.3 | XP_006719869.1 | 8 - 9 | 1765 | 106 | |
XM_011535047.2 | XP_011533349.1 | 8 - 9 | 1765 | 106 | |
XM_017020544.1 | XP_016876033.1 | 8 - 9 | 1765 | 106 | |
XM_017020545.1 | XP_016876034.1 | 8 - 9 | 1765 | 106 | |
XM_017020546.1 | XP_016876035.1 | 8 - 9 | 1053 | 106 | |
GenBank mRNA | AF467288.1 | - | 9 - 10 | 1437 | 106 |