Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NBEA
Entrez Gene ID
26960
Gene Name
neurobeachin
Gene Aliases
BCL8B, LYST2
Chromosome Location
Chr.13: 34942287 - 35672737 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204197.1 NP_001191126.1 10 - 11 1211 119
NM_015678.4 NP_056493.3 50 - 51 8241 119
XM_005266347.4 XP_005266404.2 51 - 52 8304 119
XM_005266348.4 XP_005266405.2 50 - 51 8295 119
XM_006719806.3 XP_006719869.1 49 - 50 8232 119
XM_011535046.2 XP_011533348.1 43 - 44 6435 119
XM_017020544.1 XP_016876033.1 51 - 52 8286 119
XM_017020545.1 XP_016876034.1 50 - 51 8223 119
XM_017020546.1 XP_016876035.1 51 - 52 7592 119
GenBank mRNA AB046764.1 - 17 - 18 1953 119
AF072371.1 - 4 - 5 428 119
AF467288.1 - 51 - 52 7913 119
AK091469.1 - 21 - 22 2400 119
AK122888.1 - 20 - 21 2754 119
AK127791.1 - 27 - 28 3592 119
AK294737.1 - 10 - 11 1211 119
AK297773.1 - 11 - 12 1309 119
AL137748.1 - 5 - 6 916 119
BX640760.1 - 26 - 27 3571 119

View Full Product Details