Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL27A1
Entrez Gene ID
85301
Gene Name
collagen type XXVII alpha 1
Gene Aliases
STLS
Chromosome Location
Chr.9: 114154271 - 114312516 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032888.3 NP_116277.2 19 - 20 3134 105
XM_006717308.2 XP_006717371.1 18 - 19 3148 105
XM_006717310.3 XP_006717373.1 17 - 18 947 105
XM_011519138.2 XP_011517440.1 19 - 20 2766 105
XM_011519140.1 XP_011517442.1 19 - 20 3202 105
XM_011519142.2 XP_011517444.1 19 - 20 3208 105
XM_011519143.2 XP_011517445.1 19 - 20 3206 105
XM_011519144.2 XP_011517446.1 19 - 20 3206 105
XM_011519145.2 XP_011517447.1 6 - 7 376 105
XM_017015239.1 XP_016870728.1 19 - 20 3207 105
GenBank mRNA AB058773.1 - 16 - 17 1614 105
AY149237.1 - 19 - 20 2728 105

View Full Product Details