The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032888.3 | NP_116277.2 | 20 - 21 | 3192 | 132 |
XM_006717308.2 | XP_006717371.1 | 19 - 20 | 3206 | 132 | |
XM_006717310.3 | XP_006717373.1 | 18 - 19 | 1005 | 132 | |
XM_011519138.2 | XP_011517440.1 | 20 - 21 | 2824 | 132 | |
XM_011519140.1 | XP_011517442.1 | 20 - 21 | 3260 | 132 | |
XM_011519142.2 | XP_011517444.1 | 20 - 21 | 3266 | 132 | |
XM_011519143.2 | XP_011517445.1 | 20 - 21 | 3264 | 132 | |
XM_011519144.2 | XP_011517446.1 | 20 - 21 | 3264 | 132 | |
XM_011519145.2 | XP_011517447.1 | 7 - 8 | 434 | 132 | |
XM_017015239.1 | XP_016870728.1 | 20 - 21 | 3265 | 132 | |
GenBank mRNA | AB058773.1 | - | 17 - 18 | 1672 | 132 |
AY149237.1 | - | 20 - 21 | 2786 | 132 |