Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL27A1
Entrez Gene ID
85301
Gene Name
collagen type XXVII alpha 1
Gene Aliases
STLS
Chromosome Location
Chr.9: 114154271 - 114312516 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032888.3 NP_116277.2 46 - 47 4663 113
XM_006717308.2 XP_006717371.1 45 - 46 4677 113
XM_006717310.3 XP_006717373.1 44 - 45 2476 113
XM_011519138.2 XP_011517440.1 46 - 47 4295 113
XM_011519140.1 XP_011517442.1 45 - 46 4677 113
XM_011519145.2 XP_011517447.1 33 - 34 1905 113
GenBank mRNA AB058773.1 - 43 - 44 3150 113
AY149237.1 - 46 - 47 4257 113

View Full Product Details