Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL27A1
Entrez Gene ID
85301
Gene Name
collagen type XXVII alpha 1
Gene Aliases
STLS
Chromosome Location
Chr.9: 114154271 - 114312516 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032888.3 NP_116277.2 55 - 56 5255 144
XM_006717308.2 XP_006717371.1 54 - 55 5269 144
XM_006717310.3 XP_006717373.1 53 - 54 3068 144
XM_011519138.2 XP_011517440.1 55 - 56 4887 144
XM_011519140.1 XP_011517442.1 54 - 55 5269 144
XM_011519145.2 XP_011517447.1 42 - 43 2497 144
GenBank mRNA AB058773.1 - 52 - 53 3742 144
AY149237.1 - 55 - 56 4849 144

View Full Product Details