The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032888.3 | NP_116277.2 | 58 - 59 | 5514 | 75 |
XM_006717308.2 | XP_006717371.1 | 57 - 58 | 5528 | 75 | |
XM_006717310.3 | XP_006717373.1 | 56 - 57 | 3327 | 75 | |
XM_011519138.2 | XP_011517440.1 | 58 - 59 | 5146 | 75 | |
XM_011519140.1 | XP_011517442.1 | 57 - 58 | 5528 | 75 | |
XM_011519145.2 | XP_011517447.1 | 45 - 46 | 2756 | 75 | |
GenBank mRNA | AB058773.1 | - | 55 - 56 | 4001 | 75 |
AY149237.1 | - | 58 - 59 | 5108 | 75 |