The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173600.2 | NP_775871.2 | - | 22434 | 154 |
GenBank mRNA | BG718209.1 | - | 4 - 5 | 210 | 154 |
HM801842.1 | - | - | 22434 | 154 |