The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_213589.2 | NP_998754.1 | 13 - 14 | 2000 | 60 |
XM_005246777.3 | XP_005246834.1 | 15 - 16 | 2151 | 60 | |
XM_005246779.4 | XP_005246836.1 | 15 - 16 | 3050 | 60 | |
XM_006712695.3 | XP_006712758.1 | 16 - 17 | 3131 | 60 | |
XM_011511646.2 | XP_011509948.1 | 16 - 17 | 2223 | 60 | |
XM_011511647.1 | XP_011509949.1 | 13 - 14 | 1856 | 60 | |
XM_011511648.1 | XP_011509950.1 | 13 - 14 | 1850 | 60 | |
GenBank mRNA | AB051468.1 | - | 12 - 13 | 1733 | 60 |
AJ584699.1 | - | 13 - 14 | 1994 | 60 | |
AY494951.1 | - | 13 - 14 | 1996 | 60 |