The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_213649.1 | NP_998814.1 | 2 - 3 | 227 | 132 |
NR_110305.1 | 2 - 3 | 227 | 132 | ||
XM_005269525.4 | XP_005269582.1 | 2 - 3 | 186 | 132 | |
GenBank mRNA | AW249181.1 | - | 2 - 3 | 236 | 132 |
AY269785.1 | - | 2 - 3 | 186 | 132 | |
BC063562.1 | - | 2 - 3 | 198 | 132 | |
BU959063.1 | - | 2 - 3 | 230 | 132 | |
H12949.1 | - | 2 - 3 | 182 | 132 |