The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303256.2 | NP_001290185.1 | 15 - 16 | 1896 | 111 |
NM_001303257.2 | NP_001290186.1 | 15 - 16 | 1896 | 111 | |
NM_014941.3 | NP_055756.1 | 16 - 17 | 2762 | 111 | |
XM_011530004.1 | XP_011528306.1 | 15 - 16 | 2121 | 111 | |
XM_017028667.1 | XP_016884156.1 | 15 - 16 | 2121 | 111 | |
XM_017028668.1 | XP_016884157.1 | 16 - 17 | 2216 | 111 | |
GenBank mRNA | AB020659.1 | - | 16 - 17 | 2676 | 111 |
BC019257.1 | - | 14 - 15 | 1423 | 111 | |
BC136782.1 | - | 16 - 17 | 1621 | 111 | |
CR456469.1 | - | 14 - 15 | 1422 | 111 |