Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESCO2
Entrez Gene ID
157570
Gene Name
establishment of sister chromatid cohesion N-acetyltransferase 2
Gene Aliases
2410004I17Rik, EFO2, RBS
Chromosome Location
Chr.8: 27771949 - 27812404 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF306675.1 - 1 - 2 80 110
AF306678.1 - 1 - 2 217 110

View Full Product Details