The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105562.2 | NP_001099032.1 | 14 - 15 | 2594 | 90 |
NM_006048.4 | NP_006039.2 | 13 - 14 | 2207 | 90 | |
XM_005263422.1 | XP_005263479.1 | 15 - 16 | 2571 | 90 | |
XM_011540488.2 | XP_011538790.1 | 14 - 15 | 1870 | 90 | |
XM_011540489.2 | XP_011538791.1 | 15 - 16 | 2571 | 90 | |
XM_011540490.1 | XP_011538792.1 | 15 - 16 | 2571 | 90 | |
XM_017000035.1 | XP_016855524.1 | 12 - 13 | 1657 | 90 | |
XM_017000036.1 | XP_016855525.1 | 14 - 15 | 1871 | 90 | |
GenBank mRNA | AB014584.3 | - | 13 - 14 | 2098 | 90 |
AB028839.1 | - | 13 - 14 | 2215 | 90 | |
AF043117.1 | - | 14 - 15 | 1991 | 90 | |
AF331520.1 | - | 13 - 14 | 1566 | 90 | |
AK023654.1 | - | 3 - 4 | 1321 | 90 | |
AK292444.1 | - | 13 - 14 | 1689 | 90 | |
AK304484.1 | - | 11 - 12 | 1194 | 90 | |
BC093696.1 | - | 13 - 14 | 1597 | 90 | |
BC143458.1 | - | 13 - 14 | 1597 | 90 | |
BC143460.1 | - | 12 - 13 | 1482 | 90 | |
JF289274.1 | - | 15 - 16 | 2059 | 90 |