The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105562.2 | NP_001099032.1 | 15 - 16 | 2708 | 99 |
NM_006048.4 | NP_006039.2 | 14 - 15 | 2321 | 99 | |
XM_005263422.1 | XP_005263479.1 | 16 - 17 | 2685 | 99 | |
XM_011540488.2 | XP_011538790.1 | 15 - 16 | 1984 | 99 | |
XM_011540489.2 | XP_011538791.1 | 16 - 17 | 2685 | 99 | |
XM_011540490.1 | XP_011538792.1 | 16 - 17 | 2685 | 99 | |
XM_017000035.1 | XP_016855524.1 | 13 - 14 | 1771 | 99 | |
XM_017000036.1 | XP_016855525.1 | 15 - 16 | 1985 | 99 | |
GenBank mRNA | AB014584.3 | - | 14 - 15 | 2212 | 99 |
AB028839.1 | - | 14 - 15 | 2329 | 99 | |
AF043117.1 | - | 15 - 16 | 2105 | 99 | |
AF331520.1 | - | 14 - 15 | 1680 | 99 | |
AK023654.1 | - | 4 - 5 | 1435 | 99 | |
AK292444.1 | - | 14 - 15 | 1803 | 99 | |
AK304484.1 | - | 12 - 13 | 1308 | 99 | |
BC093696.1 | - | 14 - 15 | 1711 | 99 | |
BC143458.1 | - | 14 - 15 | 1711 | 99 | |
BC143460.1 | - | 13 - 14 | 1596 | 99 | |
JF289274.1 | - | 16 - 17 | 2173 | 99 |