The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105562.2 | NP_001099032.1 | 17 - 18 | 3050 | 105 |
NM_006048.4 | NP_006039.2 | 16 - 17 | 2663 | 105 | |
XM_005263422.1 | XP_005263479.1 | 18 - 19 | 3027 | 105 | |
XM_011540488.2 | XP_011538790.1 | 17 - 18 | 2326 | 105 | |
XM_011540489.2 | XP_011538791.1 | 18 - 19 | 3027 | 105 | |
XM_011540490.1 | XP_011538792.1 | 18 - 19 | 3027 | 105 | |
XM_017000035.1 | XP_016855524.1 | 15 - 16 | 2113 | 105 | |
XM_017000036.1 | XP_016855525.1 | 17 - 18 | 2327 | 105 | |
GenBank mRNA | AB014584.3 | - | 16 - 17 | 2554 | 105 |
AB028839.1 | - | 16 - 17 | 2671 | 105 | |
AF043117.1 | - | 17 - 18 | 2447 | 105 | |
AF331520.1 | - | 16 - 17 | 2022 | 105 | |
AK292444.1 | - | 16 - 17 | 2145 | 105 | |
AK304484.1 | - | 14 - 15 | 1650 | 105 | |
BC093696.1 | - | 16 - 17 | 2053 | 105 | |
BC143458.1 | - | 16 - 17 | 2053 | 105 | |
BC143460.1 | - | 15 - 16 | 1938 | 105 | |
JF289274.1 | - | 18 - 19 | 2515 | 105 |