Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDAC6
Entrez Gene ID
10013
Gene Name
histone deacetylase 6
Gene Aliases
CPBHM, HD6, JM21, PPP1R90
Chromosome Location
Chr.X: 48801377 - 48824982 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321225.1 NP_001308154.1 27 - 28 3462 67
NM_001321226.1 NP_001308155.1 26 - 27 3872 67
NM_001321227.1 NP_001308156.1 26 - 27 3593 67
NM_001321228.1 NP_001308157.1 26 - 27 3556 67
NM_001321229.1 NP_001308158.1 26 - 27 3410 67
NM_006044.3 NP_006035.2 26 - 27 3463 67
NR_135591.1 24 - 25 3143 67
GenBank mRNA AB020708.1 - 26 - 27 3378 67
AF132609.1 - 25 - 26 3306 67
AF174499.1 - 20 - 21 2860 67
AJ011972.1 - 26 - 27 3399 67
AK001543.1 - 10 - 11 1848 67
AK095124.1 - 15 - 16 2399 67
AK122954.1 - 14 - 15 2857 67
AK127118.1 - 14 - 15 4011 67
AK131286.1 - 4 - 5 1777 67
AK302926.1 - 26 - 27 3401 67
AK303012.1 - 24 - 25 3304 67
AL137696.1 - 25 - 26 4123 67
BC013219.1 - 3 - 4 958 67
BC013737.1 - 25 - 26 3288 67
BC069243.1 - 26 - 27 3405 67

View Full Product Details