The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321225.1 | NP_001308154.1 | 6 - 7 | 555 | 94 |
NM_001321226.1 | NP_001308155.1 | 5 - 6 | 965 | 94 | |
NM_001321227.1 | NP_001308156.1 | 5 - 6 | 686 | 94 | |
NM_001321228.1 | NP_001308157.1 | 5 - 6 | 649 | 94 | |
NM_001321229.1 | NP_001308158.1 | 5 - 6 | 503 | 94 | |
NM_001321230.1 | NP_001308159.1 | 5 - 6 | 686 | 94 | |
NM_001321231.1 | NP_001308160.1 | 5 - 6 | 965 | 94 | |
NM_006044.3 | NP_006035.2 | 5 - 6 | 556 | 94 | |
NR_135591.1 | 4 - 5 | 429 | 94 | ||
NR_135592.1 | 4 - 5 | 563 | 94 | ||
NR_135593.1 | 4 - 5 | 563 | 94 | ||
XM_017029187.1 | XP_016884676.1 | 5 - 6 | 478 | 94 | |
GenBank mRNA | AB020708.1 | - | 5 - 6 | 471 | 94 |
AF132609.1 | - | 4 - 5 | 399 | 94 | |
AJ011972.1 | - | 5 - 6 | 492 | 94 | |
AK024083.1 | - | 5 - 6 | 498 | 94 | |
AK295884.1 | - | 4 - 5 | 416 | 94 | |
AK302926.1 | - | 5 - 6 | 494 | 94 | |
AK303012.1 | - | 3 - 4 | 397 | 94 | |
AL137696.1 | - | 5 - 6 | 520 | 94 | |
BC005872.1 | - | 5 - 6 | 925 | 94 | |
BC011498.1 | - | 5 - 6 | 512 | 94 | |
BC013737.1 | - | 4 - 5 | 381 | 94 | |
BC069243.1 | - | 5 - 6 | 498 | 94 | |
BT006649.1 | - | 4 - 5 | 399 | 94 | |
DA191923.1 | - | 4 - 5 | 372 | 94 |