The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014421.2 | NP_055236.1 | 1 - 2 | 932 | 80 |
GenBank mRNA | AB033208.1 | - | 1 - 2 | 449 | 80 |
AB033941.1 | - | 1 - 2 | 449 | 80 | |
AF177395.1 | - | 1 - 2 | 944 | 80 | |
AK312695.1 | - | 1 - 2 | 458 | 80 | |
AY358414.1 | - | 1 - 2 | 437 | 80 | |
BC075077.2 | - | 1 - 2 | 261 | 80 | |
BC075078.2 | - | 1 - 2 | 261 | 80 | |
BC126328.1 | - | 1 - 2 | 533 | 80 | |
BC126330.1 | - | 1 - 2 | 533 | 80 |