Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRX
Entrez Gene ID
57716
Gene Name
periaxin
Gene Aliases
CMT4F
Chromosome Location
Chr.19: 40393762 - 40414718 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020956.2 NP_066007.1 4 - 5 298 79
NM_181882.2 NP_870998.2 4 - 5 298 79
XM_011527171.2 XP_011525473.1 4 - 5 290 79
GenBank mRNA AF321191.1 - 4 - 5 298 79
AF321192.1 - 4 - 5 298 79
BC067266.1 - 4 - 5 300 79

View Full Product Details