Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PER3
Entrez Gene ID
8863
Gene Name
period circadian clock 3
Gene Aliases
FASPS3, GIG13
Chromosome Location
Chr.1: 7784285 - 7845181 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289861.1 NP_001276790.1 21 - 22 3610 89
NM_001289862.1 NP_001276791.1 20 - 21 3667 89
NM_001289863.1 NP_001276792.1 21 - 22 3589 89
NM_001289864.1 NP_001276793.1 20 - 21 3725 89
NM_016831.2 NP_058515.1 20 - 21 3640 89
XM_005263524.3 XP_005263581.1 19 - 20 3542 89
XM_011542385.2 XP_011540687.1 19 - 20 3539 89
XM_011542390.2 XP_011540692.1 19 - 20 3518 89
XM_017002723.1 XP_016858212.1 19 - 20 3539 89
XM_017002724.1 XP_016858213.1 19 - 20 3536 89
XM_017002725.1 XP_016858214.1 19 - 20 3521 89
XM_017002726.1 XP_016858215.1 19 - 20 3518 89
XM_017002727.1 XP_016858216.1 21 - 22 3861 89
XM_017002728.1 XP_016858217.1 19 - 20 3603 89
XM_017002729.1 XP_016858218.1 17 - 18 3043 89
XM_017002730.1 XP_016858219.1 19 - 20 3600 89
XM_017002731.1 XP_016858220.1 19 - 20 3579 89
XM_017002732.1 XP_016858221.1 18 - 19 3631 89
XM_017002733.1 XP_016858222.1 21 - 22 3768 89
XM_017002734.1 XP_016858223.1 22 - 23 3910 89
XM_017002735.1 XP_016858224.1 21 - 22 3765 89
XM_017002736.1 XP_016858225.1 22 - 23 3889 89
XM_017002737.1 XP_016858226.1 16 - 17 3026 89
XM_017002738.1 XP_016858227.1 16 - 17 3029 89
GenBank mRNA AB047686.1 - 19 - 20 3627 89
AK299123.1 - 4 - 5 596 89
AK307760.1 - 20 - 21 3465 89
AK308303.1 - 21 - 22 3660 89
AL157954.1 - 19 - 20 3551 89
AY493418.1 - 19 - 20 3447 89
BC111010.1 - 4 - 5 854 89
BC144137.1 - 19 - 20 3629 89
BC146781.1 - 19 - 20 3627 89

View Full Product Details