The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003652.3 | NP_001003652.1 | 8 - 9 | 1400 | 95 |
NM_001135937.2 | NP_001129409.1 | 7 - 8 | 1310 | 95 | |
NM_005901.5 | NP_005892.1 | 8 - 9 | 1292 | 95 | |
XM_005258259.3 | XP_005258316.1 | 9 - 10 | 1533 | 95 | |
XM_006722451.3 | XP_006722514.1 | 9 - 10 | 1545 | 95 | |
XM_011525984.2 | XP_011524286.1 | 8 - 9 | 889 | 95 | |
XM_011525985.2 | XP_011524287.1 | 10 - 11 | 1592 | 95 | |
XM_017025745.1 | XP_016881234.1 | 10 - 11 | 1631 | 95 | |
XM_017025746.1 | XP_016881235.1 | 8 - 9 | 1443 | 95 | |
XM_017025747.1 | XP_016881236.1 | 9 - 10 | 1518 | 95 | |
XM_017025748.1 | XP_016881237.1 | 10 - 11 | 1604 | 95 | |
XM_017025750.1 | XP_016881239.1 | 9 - 10 | 1493 | 95 | |
GenBank mRNA | AF027964.1 | - | 8 - 9 | 1106 | 95 |
AK299218.1 | - | 7 - 8 | 1006 | 95 | |
AK312963.1 | - | 7 - 8 | 1129 | 95 | |
BC014840.1 | - | 8 - 9 | 1248 | 95 | |
BC025699.1 | - | 8 - 9 | 1106 | 95 | |
BT007422.1 | - | 7 - 8 | 1000 | 95 | |
U59911.1 | - | 8 - 9 | 1099 | 95 | |
U65019.1 | - | 7 - 8 | 1036 | 95 | |
U68018.1 | - | 7 - 8 | 1055 | 95 |