The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004124.2 | NP_004115.1 | 5 - 6 | 385 | 135 |
GenBank mRNA | AB001106.1 | - | 5 - 6 | 383 | 135 |
AK311753.1 | - | 5 - 6 | 319 | 135 | |
BC005359.1 | - | 5 - 6 | 339 | 135 | |
BI547506.1 | - | 5 - 6 | 395 | 135 | |
BT006784.1 | - | 4 - 5 | 286 | 135 | |
CR542113.1 | - | 4 - 5 | 286 | 135 | |
M86492.1 | - | 5 - 6 | 326 | 135 |