The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004455.2 | NP_004446.2 | 4 - 5 | 1968 | 132 |
XM_005245779.3 | XP_005245836.1 | 3 - 4 | 1891 | 132 | |
XM_017000650.1 | XP_016856139.1 | 4 - 5 | 1237 | 132 | |
GenBank mRNA | BC065528.1 | - | 4 - 5 | 1814 | 132 |
U67191.1 | - | 4 - 5 | 1967 | 132 |