Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH2D3A
Entrez Gene ID
10045
Gene Name
SH2 domain containing 3A
Gene Aliases
NSP1
Chromosome Location
Chr.19: 6752160 - 6767513 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005490.2 NP_005481.2 1 - 2 133 99
XM_005259467.1 XP_005259524.1 1 - 2 83 99
XM_006722602.1 XP_006722665.1 1 - 2 83 99
XM_017026123.1 XP_016881612.1 1 - 2 83 99
XM_017026124.1 XP_016881613.1 1 - 2 83 99
GenBank mRNA AF124249.1 - 1 - 2 90 99
AK290293.1 - 1 - 2 209 99
AY358406.1 - 1 - 2 90 99
BC006281.2 - 1 - 2 70 99

View Full Product Details